Learn More
3M™ Light Vision™ and Light Vision™ 2 Eyewear LED Eyewear
Catalog No. 19040065 Shop All 3M Products
Change view
Click to view available options
Includes:
Light Vision2
Magnification Power:
+1.50
Catalog No. Includes Magnification Power
19-040-065 Light Vision2 +1.50
1 options
This item is not returnable. View return policy
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings and limitations accompanying each product.
Catalog No. 19-040-065 Supplier 3M™ Supplier No. 7000127506
This item is not returnable. View return policy
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings and limitations accompanying each product.
Includes
Light Vision and Light Vision 2 glasses supplied with lanyard and storage bag. Light Vision OTG supplied with storage bag.
Industrial eyewear designed with integral hands-free lighting; work safely in dark environments
  • Dual hands-free temple LED lights swivel and adjust for bright directional beams
  • Wraparound design provides wide coverage to protect against particulates
  • Impact-resistant, clear polycarbonate lens with antifog coating absorbs 99.9% UV
  • Soft temples and nose pads for extended wear comfort
  • Light Vision 2 glasses feature adjustable temple lengths
  • Over-the-glass style fits over prescription glasses; features adjustable length temples and Secure-Grip™ temple tips
  • Light Vision 2 available with bifocal readers in three strengths for workers who have difficulty reading small print or are engaged in small detail work
  • Batteries provide up to 50 hours of continuous use
  • 3M Catalog# 11356-00000-10, 11476-00000-10, 11477-00000-10, 11478-00000-10, 11479-00000-10
Compliance
Meets ANSI™ Z87.1-2010 standards. LightVision 2 also meets CSA Z94.3-2007 standards.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Meets ANSI Z87.1-2010 standards. LightVision 2 also meets CSA Z94.3-2007 standards.
Lens Tint Clear
Frame Color Gray
Includes Light Vision2
Magnification Power +1.50
Temple Color Black
Lens Material Polycarbonate