Learn More
MSC 3M LINERED VINYL TAPE
Catalog No. 5000802886
Encompass
This floor and aisle marking tape is designed for light-duty traffic. It is 3 in. wide, 108 ft long, and 5.2 mil thick, made of rubber material.
  • Available in a wide variety of vivid colors for diverse applications
  • Unique stretch properties enable tape to remain stretched to conform to uneven surfaces without lifting and retracting
  • One-piece, clean removal from many surfaces which helps reduce clean-up and labor costs
  • Rubber adhesive provides excellent holding strength on many surfaces
  • Good abrasion and solvent resistance for long service life
Catalog No. 50-008-02886 Supplier MSC Supplier No. 00048827
This floor and aisle marking tape is designed for light-duty traffic. It is 3 in. wide, 108 ft long, and 5.2 mil thick, made of rubber material.
  • Available in a wide variety of vivid colors for diverse applications
  • Unique stretch properties enable tape to remain stretched to conform to uneven surfaces without lifting and retracting
  • One-piece, clean removal from many surfaces which helps reduce clean-up and labor costs
  • Rubber adhesive provides excellent holding strength on many surfaces
  • Good abrasion and solvent resistance for long service life