Vinyl Tape 471+ is a conformable tape designed for fine line and critical paint masking. This durable tape is flexible and removes cleanly even after bake cycles, without adhesive build-up or ghosting. It ensures exceptional paint lines in various painting applications.
- Excels in masking applications to create sharp, fine paint lines for striping and complex designs.
- Semi-translucent indigo backing provides visual see-through for accurate alignment.
- Clean removal without ghosting, even after a 30-minute bake cycle at temperatures up to 121°C.
- Unique stretch properties enable the tape to conform to uneven surfaces without lifting.
- Rubber adhesive provides quick stick with excellent holding strength on most surfaces.