Learn More
MSC 3M LINERED VINYL TAPE
Catalog No. 5000803563
Encompass
This floor and aisle marking tape is designed for light-duty traffic and features a non anti-slip surface. It is blue in color and constructed from vinyl for both the tape material and the adhesive, making it suitable for aisle marking applications.
  • Designed for light-duty traffic
  • Non anti-slip surface
  • Made from vinyl material and adhesive
  • Available in a wide variety of vivid colors
  • Unique stretch properties allow conformity to uneven surfaces
  • One-piece, clean removal from many surfaces
  • Rubber adhesive provides excellent holding strength
  • Good abrasion and solvent resistance for long service life
  • Dimensions: 1.5 in wide, 108 ft long, 5.2 mil thick
  • Pattern: solid
Catalog No. 50-008-03563 Supplier MSC Supplier No. 65368839
This floor and aisle marking tape is designed for light-duty traffic and features a non anti-slip surface. It is blue in color and constructed from vinyl for both the tape material and the adhesive, making it suitable for aisle marking applications.
  • Designed for light-duty traffic
  • Non anti-slip surface
  • Made from vinyl material and adhesive
  • Available in a wide variety of vivid colors
  • Unique stretch properties allow conformity to uneven surfaces
  • One-piece, clean removal from many surfaces
  • Rubber adhesive provides excellent holding strength
  • Good abrasion and solvent resistance for long service life
  • Dimensions: 1.5 in wide, 108 ft long, 5.2 mil thick
  • Pattern: solid