Learn More
Enterprise Technology Solutions 3M Marking Tape, Vinyl, 1"x108', Yellow
SDP
Catalog No. 5000828501
Encompass_Preferred
Marking Tape is ideal for many color-coding, abrasion protection, decoration, sealing, patching, splicing, wrapping and general-purpose applications. Excellent for identifying caution areas. Tape resists many common solvents except ketones, chlorinated hydrocarbons and esters found in lacquer thinner, degreasers, paint strippers and more.
Catalog No. 50-008-28501 Supplier Enterprise Technology Solutions Supplier No. MMM4711YE
Marking Tape is ideal for many color-coding, abrasion protection, decoration, sealing, patching, splicing, wrapping and general-purpose applications. Excellent for identifying caution areas. Tape resists many common solvents except ketones, chlorinated hydrocarbons and esters found in lacquer thinner, degreasers, paint strippers and more.