Learn More
Enterprise Technology Solutions 3M Masking Tape,Industrial,18mmx55m,Green
SDP
Catalog No. 5000828833
Encompass_Preferred
High-Performance Green Masking Tape 401Plus delivers sharp, clean paint lines and is ideal for high-temperature paint-baking, fascia painting and powder-coating applications. This high-strength, crepe-paper backing conforms to irregular surfaces, resists bleed-through from most industrial paint systems and improves visibility during the paint-masking process to help create sharp paint lines. It sticks instantly to most surfaces, including metal, rubber, glass and plastic as well as EPDM type rubber moldings. Masking tape withstands surface temperatures up to 250 degrees Fahrenheit/121 degrees Celsius for 30 minutes in paint-bake operations. This crepe paper tape is ideal for critical paint-masking applications in automotive, specialty vehicle and industrial markets where a strong hold and high-temp performance are required. Remove the proprietary, solvent-free adhesive cleanly in one piece with no adhesive residue. Tape is easy to tear yet strong enough to resist slivering.
Catalog No. 50-008-28833 Supplier Enterprise Technology Solutions Supplier No. MMM40118X55
High-Performance Green Masking Tape 401Plus delivers sharp, clean paint lines and is ideal for high-temperature paint-baking, fascia painting and powder-coating applications. This high-strength, crepe-paper backing conforms to irregular surfaces, resists bleed-through from most industrial paint systems and improves visibility during the paint-masking process to help create sharp paint lines. It sticks instantly to most surfaces, including metal, rubber, glass and plastic as well as EPDM type rubber moldings. Masking tape withstands surface temperatures up to 250 degrees Fahrenheit/121 degrees Celsius for 30 minutes in paint-bake operations. This crepe paper tape is ideal for critical paint-masking applications in automotive, specialty vehicle and industrial markets where a strong hold and high-temp performance are required. Remove the proprietary, solvent-free adhesive cleanly in one piece with no adhesive residue. Tape is easy to tear yet strong enough to resist slivering.