Learn More
3M™ Maxim 2X2 Air Flow Goggles
Catalog No. 19065419 Shop All 3M Products
This item is not returnable. View return policy
Catalog No. 19-065-419 Supplier 3M™ Supplier No. 7000127226
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Designed for tactical use, the Maxim 2x2 protective, polycarbonate air flow goggle is both stylish and durable. Featuring flexible fit temples, a vented anti-fog seal and an elastic strap for a secure fit, it maximizes visual comfort and clarity.
The Maxim 2X2 air flow safety goggle is a preferred solution for people in tactical situations. The lightweight safety goggle was designed to help protect from sun, wind, dust and flying debris, while incorporating expansive top and bottom venting to reduce fog and heat. Its dual-lens design enhances sightline and field of vision.
  • Anti-fog lens.
  • Nylon frame
  • Elastic strap
  • Meets the High Impact Requirements of ANSI Z87.1-2010
  • 10 safety goggles per case
  • Vented seals and anti-fog coating keeps your vision clear
  • Multi-protection goggles help protect from wind, dust, and flying debris
  • Choice of elastic strap for a secure fit or flexible fit temples
  • Available in two lens tints – clear and gray
  • Polycarbonate lenses absorb 99.9% UVA and UVB up to 380 nm
  • 3M Catalog# 40698-00000 , 3M Catalog# 40699-00000
TRUSTED_SUSTAINABILITY
Specifications
Lens Tint Clear
Product Type Safety goggles
Includes Black Strap