Description
- Lightweight frame with polymetric materials ensure comfort at cheeks, forehead, and nose
- Duosoft shroud for all-day comfort
- Indirect ventilation for improved circulation, comfort and reduced fogging
- Fits over most prescription eyewear
- Anti-fog coated lens
Compliance
Meets ANSI Z87.1-2015 standards.
Specifications
Specifications
| Certifications/Compliance | ANSI Z87.1-2015 |
| Lens Tint | Clear |
| Product Type | Splash Goggle |
| Ventilation Details | Indirect vent |