Learn More
3M™ Model 200 Replacement Pods
Catalog No. 19048620 Shop All 3M Products
Change view
Click to view available options
Noise Reduction Rating (NRR):
17 dB
Catalog No. Noise Reduction Rating (NRR)
19-048-620 17 dB
1 options
Catalog No. 19-048-620 Supplier 3M™ Supplier No. 7000127665
Add to Cart
Add to Cart
Replacement Pods for E-A-R Caps Model 200 Hearing Protector (350-2100 and 321-2101).
  • Replacement foam caps for E-A-R™ Caps Model 200 Hearing Protector 350-2100 and 321-2101
  • Soft foam replacement caps for use only with E-A-R™ Caps banded hearing protector
    • Not for use without band
The NRR may overestimate the hearing protection provided during typical use. 3M recommends reducing the NRR by 50% for estimating the amount of noise reduction provided.
Compliance
3M™ ear plugs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
TRUSTED_SUSTAINABILITY
Specifications
ANSI S3.1974 Yes
CSA Z94.2 Class AL
Size Universal
For Use With (Application) Assembly, Blasting, Cleaning, Demolition, Electrical, Facility Maintenance, Grinding, Machine Operations, Painting, Sanding, Welding
High Visibility Yes
Model 321-2103
Plug Material Foam
Plug Color Yellow
Noise Reduction Rating (NRR) 17 dB
Plug Shape Pod
Disposable Single-use
Metal Detectable No
Dielectric No