Learn More
3M™ Modul-R Safety Goggle Clear Anti Fog Lens with Chin Protector
Catalog No. 19065422 Shop All 3M Products
Change view
Click to view available options
Lens Coating:
Anti-fog
Catalog No. Lens Coating
19-065-422 Anti-fog
1 options
This item is not returnable. View return policy
Catalog No. 19-065-422 Supplier 3M™ Supplier No. 7000030008
This item is not returnable. View return policy
Modul-R protective splash safety goggles offer a wider viewing area, strong polycarbonate lens construction and a flexible strap to provide full profile protection. Splash shield has an adjustable venting system to minimize fog.
Modul-R splash goggles deliver reliable performance and protection in a snap. It is equipped with a splash shield. Its strap pivots for added adjustability, and includes a low profile with an extended field of vision
  • Low profile lens with an extended field of vision
  • Strap pivots for added adjustability
  • Splash shield has an adjustable venting system to help minimize fog and splash
  • 3M Catalog# 40658-00000-10
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog
Certifications/Compliance ANSI Z87.1-2003
Lens Tint Clear
Product Type Safety Goggles
Includes Chin protector; Flexible strap; Splash shield with adjustable venting system
Model 40658-00000-10
Material Polycarbonate