Learn More
Enterprise Technology Solutions 3M Multipurpose Adhesive Spray, Aerosol, 13.75 oz, Clear
SDP
Catalog No. 5000830106
Encompass_Preferred
Scotch Super 77 Multipurpose Adhesive is an extremely versatile, fast-drying spray adhesive that bonds a wide range of lightweight materials. Its fast, aggressive tack and low soak-in make it ideal for a broad range of projects. Use it for bonding paper, cardboard, fabric, cushion foam, plastic, metal, wood and much more. Spray adhesive dries quickly and is transparent when dried. Adhesive contains 1,1-Difluoroethane (DFE).
Catalog No. 50-008-30106 Supplier Enterprise Technology Solutions Supplier No. MMM77L
Scotch Super 77 Multipurpose Adhesive is an extremely versatile, fast-drying spray adhesive that bonds a wide range of lightweight materials. Its fast, aggressive tack and low soak-in make it ideal for a broad range of projects. Use it for bonding paper, cardboard, fabric, cushion foam, plastic, metal, wood and much more. Spray adhesive dries quickly and is transparent when dried. Adhesive contains 1,1-Difluoroethane (DFE).