Learn More
Enterprise Technology Solutions 3M Multipurpose Respirator Replacement Cartridges, 2/PK, PK
SDP
Catalog No. 5000828178
Encompass_Preferred
Protection Replacement Cartridges are designed for use with 3M 6000 or 7500 Series half-facepiece respirators (sold separately) for professional removal of certain mold, lead and sprays from coatings and sealants. Cartridges are NIOSH approved and offer at least 99.7 percent efficiency against solid and liquid aerosols, including oils. Easy-to-use replacement cartridges are a great choice in respiratory protection for the professional. Cartridges and respirators are not recommended for Do-it-Yourself applications.
Catalog No. 50-008-28178 Supplier Enterprise Technology Solutions Supplier No. MMM60923HB1C
Protection Replacement Cartridges are designed for use with 3M 6000 or 7500 Series half-facepiece respirators (sold separately) for professional removal of certain mold, lead and sprays from coatings and sealants. Cartridges are NIOSH approved and offer at least 99.7 percent efficiency against solid and liquid aerosols, including oils. Easy-to-use replacement cartridges are a great choice in respiratory protection for the professional. Cartridges and respirators are not recommended for Do-it-Yourself applications.