Learn More
3M™ N95 and P95 Particulate Filters (5000/6000 Series)
Catalog No. 189994547 Shop All 3M Products
Change view
Click to view available options
Product Type:
N95 Particulate Filter
P95 Particulate Filter
For Use With (Application):
Assembly, cleaning, demolition, facility maintenance, grinding, machine operation, painting, sanding, welding
Cleaning, demolition, facility maintenance, grinding, painting, sanding, welding
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type For Use With (Application)
18-999-4547 N95 Particulate Filter Assembly, cleaning, demolition, facility maintenance, grinding, machine operation, painting, sanding, welding
19-002-717 P95 Particulate Filter Cleaning, demolition, facility maintenance, grinding, painting, sanding, welding
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 18-999-4547 Supplier 3M™ Supplier No. 7000002008
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Requires
Filter retainer 17-673B for use with 5000 Series Respirators and 6000 Series Cartridges.
NIOSH approved for environments containing certain non-oil based particles. Use with 3M™ Respirators 5000 Series or 3M™ Cartridges 6000 Series or 3M™ Filter Adapter 603, and 3M™ Filter Retainer 501.
The 3M™ Particulate Filter 5N11, N95 helps provide respiratory protection in environments containing certain non-oil based particles. When properly fitted, use with appropriate cartridges for various applications including spray painting, petrochemical and chemical manufacturing, and pesticide spraying for concentrations up to 10 times the Permissible Exposure Limit (PEL) with half facepieces, or 50 times PEL with quantitatively fit tested full facepieces. Filter protects against: 1) solids such as those from processing minerals, coal, iron ore, cotton, flour, and certain other substances; 2) liquids or oil based particles from sprays that do not emit harmful vapors; and 3) metal fumes produced from welding, brazing, cutting, and other operations involving heating of metals.NIOSH (National Institute for Occupational Safety and Health) approved for environments containing certain non-oil based particles. This filter has been rated N95, which means it filters at least 95% of airborne particles, but is not resistant to oil.Breathing hazardous particles can pose a risk to your health. NIOSH, a Federal government regulatory agency, has tested and approved the 3M™ Particulate Filter 5N11 to help reduce exposure to certain particles.
  • NIOSH approved for environments containing certain non-oil-based particles
  • Replaceable particle filter for cost savings
  • Wide range of applications reduces inventory needs and training requirements
  • Designed for use with 3M™ Respirators 5000 Series or 3M™ Cartridges 6000 Series or 3M™ Filter Adapter 603, and 3M™ Filter Retainer 501
  • 3M Catalog# 5N11 , 3M Catalog# 5P71
TRUSTED_SUSTAINABILITY
Specifications
Cartridge Type Filter Cartridge
Certifications/Compliance NIOSH
Product Type N95 Particulate Filter
For Use With (Application) Assembly, cleaning, demolition, facility maintenance, grinding, machine operation, painting, sanding, welding
For Use With (Equipment) 3M™ respirator 5000 series or 3M™ cartridge 6000 series or 3M™ filter adapter 603 and 3M™ filter retainer 501
Model 5N11
Length (English) 4.2 in.
Length (Metric) 10.6 cm
Height (English) 3.4 in.
Height (Metric) 8.6 cm