Learn More
3M™ Nassau RAVE Safety Glasses
Catalog No. 19038495 Shop All 3M Products
Change view
Click to view available options
Black Frame and Green Shade 5.0 Lens
Catalog No.
19-038-495 Black Frame and Green Shade 5.0 Lens
1 options
This item is not returnable. View return policy
Catalog No. 19-038-495 Supplier 3M™ Supplier No. 7010045666
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Nassau Rave protective eyewear features a distinctive unisex design with adjustable lens angles and temple lengths. Minimizes fogging with vertically vented browgard. Its wraparound lens helps provide front and side protection.
Nassau Rave protective eyewear features a distinctive unisex design with adjustable lens angles and temple lengths. Minimizes fogging with vertically vented browgard. Its wraparound lens helps provide front and side protection.
  • Shade 3.0 IR lens
  • Black frame
  • 3M™ DX™ anti-fog hard coat protects from fog, scratching, and many chemicals
  • Polycarbonate lens absorbs 99.9% UV
  • Meets the requirements of CSA Z94.3-2007 and the High Impact Requirements of ANSI Z87.1-2003
  • 20 safety glasses per case
  • 3M Catalog# 14459-00000-20 , 3M Catalog# 14460-00000-20
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ANSI Z87.1, CAN / CSA-Z94.3
Description Green Shade
Lens Tint Green
Frame Color Black
Model 14460-00000-20