Learn More
3M™ Nitrous™ CCS Eyewear
Catalog No. 17827308 Shop All 3M Products
Change view
Click to view available options
Lens Tint:
Clear
Catalog No. Lens Tint
17-827-308 Clear
1 options
This item is not returnable. View return policy
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Catalog No. 17-827-308 Supplier 3M™ Supplier No. 118010000020
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Features an attached, corded earplug—untangled and ready for use
  • Lightweight, contemporary design
  • Antifog-coated polycarbonate lenses absorb 99.9% UV
  • Comfortable, soft temple tips
  • Corded Control System (CCS) compatible with a variety of vinyl-corded earplugs; can also be used as a lanyard
Compliance
Meets ANSI™ Z87. 1-2010 standards.
TRUSTED_SUSTAINABILITY
Specifications
Lens Tint Clear
Frame Color Black
No. per Case 20/Cs.
Lens Material Polycarbonate