Learn More
3M™ Nitrous Protective Bronze Anti-Fog Lens
Catalog No. 19176320 Shop All 3M Products
Change view
Click to view available options
Lens Coating:
Anti-fog
Catalog No. Lens Coating
19-176-320 Anti-fog
1 options
Catalog No. 19-176-320 Supplier 3M™ Supplier No. 7010340619
Sleek and edgy Nitrous unisex protective eyewear merges style with safety, fitted with soft rubber temples and nose bridge, and an anti-fog, polycarbonate, scratch-resistant lens. Comfortable, contoured safety glasses for indoor or outdoor use.
  • Approved 3M replacement part
  • Designed for use with the 3M Ultimate FX Full Facepiece Reusable Respirators (sold separately)
  • Maintains original performance standards of facepiece
  • Weighs only 82 ounces
  • Replaceable part can potentially result in respiratory equipment cost savings
  • 3M Catalog# 11712-00000-20
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog
Description Lens; Includes lanyard and microfiber bag
Product Type Replacement Lens
Model 11712-00000-20