Learn More
3M™ Nuvo™ Readers Eyewear
Catalog No. 19123503 Shop All 3M Products
Change view
Click to view available options
Lens Tint:
Clear
Gray
Magnification Power:
+1.50
+2.00
+2.50
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint Magnification Power
19-123-503 Clear +2.00
19-123-502 Clear +1.50
19-123-504 Clear +2.50
19-130-0820 Gray +2.00
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Catalog No. 19-123-503 Supplier 3M™ Supplier No. 7000052798
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Bifocal readers with three magnification diopters
Designed for workers who have difficulty reading small print or who are engaged in fine detail work.
  • Classic look of a dual lens but with tough, lightweight one-piece lens
  • Lens with anti-fog coating absorbs 99.9% UV
  • Gray, adjustable length temples with soft tips for comfortable fit
  • Soft nosepiece
  • Integral sideshields provide profile protection
  • Cushioned brow bar
  • 3M Catalog# 11434-00000-20, 11435-00000-20, 11436-00000-20, 11500-00000-20, 11501-00000-20
Compliance
Meets ANSI Z87.1-2015 standards.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog, Scratch-resistant
Certifications/Compliance ANSI Z87.1-2015
Lens Tint Clear
Product Type Bifocal Readers
For Use With (Application) Designed for workers who have difficulty reading small print or who are engaged in fine detail work
Magnification Power +2.00
Model 11435-00000-20
UV Protection UV400