Learn More
3M™ Nuvo Translucent Eyewear
Catalog No. 19121255 Shop All 3M Products
Change view
Click to view available options
Lens Coating:
Anti-fog
Catalog No. Lens Coating
19-121-255 Anti-fog
1 options
This item is not returnable. View return policy
Catalog No. 19-121-255 Supplier 3M™ Supplier No. 7000127497
Add to Cart
Add to Cart
This item is not returnable. View return policy
Lightweight, retro-styled Nuvo protective eyewear features adjustable temples, an integral brow bar and soft nosepiece. Contoured frame and sideshields offer additional comfort.
3M™ Nuvo™ Protective Eyewear combines a distinct, nostalgic look with reliable eye protection that includes an integral brow bar and side shields for added coverage. The glasses appear to have a dual lens to make them look like standard glasses, but the lens is really a one-piece design for extra durability. The impact-resistant polycarbonate lens absorbs 99.9% UV rays. To enhance comfort, these safety glasses come with a soft nose bridge to cushion a high contact point. These safety glasses are recommended for the following tasks: chipping, chiseling, drilling, grinding, machining, masonry, pouring/casting, power fastening, riveting, sanding and sawing. Industries in which these safety glasses are commonly used include aviation, construction, food and beverage, general manufacturing, mining, oil and gas, primary metals and transportation. Nuvo™ safety glasses meet the requirements of CSA Z94.3-2007 and the high impact requirements of ANSI Z87.1-2003.Eye protection is important to safeguard your eyes and maintain healthy vision. According to the Bureau of Labor Statistics, nearly three out of every five workers who suffered eye injuries were not wearing eye protection at the time of the accident. Wearing the correct safety eyewear for the task and for the unique needs of the worker is important. 3M™ recognizes the need to design comfortable and attractive eyewear, increasing the usage of safety glasses on the job. That’s why all 3M™ protective eyewear combines safety features with comfort enhancements.
  • “Retro” styling gives the glasses a nostalgic look
  • Integral brow bar and side shields offer additional protection
  • Dual lens profile in a one-piece lens design combines style with durability
  • Soft nose bridge cushions a high contact point
  • Polycarbonate lenses absorb 999% UV
  • 3M Catalog# 11411-00000-20
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog
Certifications/Compliance Meets ANSI Z87.1 and CSA Z94.3 standards
Frame Color Translucent
Model 11411-00000-20