Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Organic Vapor Service Life Indicator Cartridges

Catalog No. 19070351 Shop All 3M Products
Change view
Click to view available options
Filter Type:
P100
Color:
Gray
Pink
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Filter Type Color
19-070-351 P100 Pink
19-070-350 - Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-070-351 Supplier 3M™ Supplier No. 7100074213

Cartridges feature a unique end-of-service-life indicator (ESLI) to help users determine when to change their cartridges in appropriate environments.

  • End-of-service-life indicator (ESLI) complements current respirator change-out schedule
  • ESLI is located inside cartridge, next to activated carbon
  • Particulate filter is permanently attached to cartridge
  • Spring-loaded filter cover
  • Designed to indicate service life based on individual exposure and respiratory use patterns

Working:

  • Organic vapors travel through the cartridge and also absorbed into the ESLI
  • Clear cartridge wall allows to monitor developing indicator bar
  • When the cartridge is exposed to specific vapor concentrations, change in the indicator is noticed

Usage:

  • When used properly in appropriate environments, indicator bar will develop to help determine remaining cartridge service life
  • Change your cartridge at the normally scheduled interval or when ESLI indicates, whichever occurs first

Certifications

NIOSH approved

TRUSTED_SUSTAINABILITY

Specifications

Cartridge Type Organic Vapors
Certifications (NIOSH) NIOSH-approved
Connection Type Bayonet
Color Pink
Filter Type P100
Product Type Service Life Indicator Cartridge
For Use With (Equipment) 3M™ 6000, 6500, 7000 and FF-400 Series Facepieces
Model 60921I
Quantity 1 Pair
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.