Learn More
Enterprise Technology Solutions 3M Painter's Tape, Multi Surface, 0.94"x60 Yds, 6/PK, Blue
SDP
Catalog No. 5000828784
Encompass_Preferred
ScotchBlue Painter's Tape Original Multi-Surface removes cleanly for up to 14 days and does not cause surface damage, even when exposed to direct sunlight. The medium-adhesion tape delivers sharp paint lines and can be used on a variety of surfaces, such as walls, trim, glass and metal. It easily conforms and sticks to the surface with consistent surface contact and secure holding power. The adhesive-transfer resistance from the acrylic adhesive, increased light and ultraviolet stability mean the tape stays intact when removed to reduce labor and rework. Solvent-resistance and moisture-resistance resist paint bleed-through, and adhesive doesn't soften or transfer when tape is wet. The tape is easy to tear and use with a smooth unwind.
Catalog No. 50-008-28784 Supplier Enterprise Technology Solutions Supplier No. MMM209024EP6
ScotchBlue Painter's Tape Original Multi-Surface removes cleanly for up to 14 days and does not cause surface damage, even when exposed to direct sunlight. The medium-adhesion tape delivers sharp paint lines and can be used on a variety of surfaces, such as walls, trim, glass and metal. It easily conforms and sticks to the surface with consistent surface contact and secure holding power. The adhesive-transfer resistance from the acrylic adhesive, increased light and ultraviolet stability mean the tape stays intact when removed to reduce labor and rework. Solvent-resistance and moisture-resistance resist paint bleed-through, and adhesive doesn't soften or transfer when tape is wet. The tape is easy to tear and use with a smooth unwind.