Learn More
Enterprise Technology Solutions 3M PainterTape,ExtraStrength,f/RoughSurfaces,1.41"x60.1yd,Green
SDP
Catalog No. 5000828952
Encompass_Preferred
Rough-Surface Painter's Tape uses extra-strong adhesive to deliver clean paint lines on rough, non-damageable surfaces. Use on brick, concrete, stucco, rough wood and more for protection you need to get the job done. For best results, apply to clean and dry surfaces, press down firmly and let tape set for 30-60 minutes before painting. Lift slowly and peel at a 45-degree angle for effective removal. Extra-strong tape holds to surfaces for up to five days and removes easily without leaving behind any sticky residues. Use with or on lacquer coatings is recommended.
Catalog No. 50-008-28952 Supplier Enterprise Technology Solutions Supplier No. MMM206036AP
Rough-Surface Painter's Tape uses extra-strong adhesive to deliver clean paint lines on rough, non-damageable surfaces. Use on brick, concrete, stucco, rough wood and more for protection you need to get the job done. For best results, apply to clean and dry surfaces, press down firmly and let tape set for 30-60 minutes before painting. Lift slowly and peel at a 45-degree angle for effective removal. Extra-strong tape holds to surfaces for up to five days and removes easily without leaving behind any sticky residues. Use with or on lacquer coatings is recommended.