Learn More
Enterprise Technology Solutions 3M Paper Masking Tape, 2"x60 Yds, 3" Core, 48mm, 6.5 mil
SDP
Catalog No. 5000829172
Encompass_Preferred
High-performance paper masking tape produces sharp paint lines in medium-temperature, paint bake operations and provides clean removal, even on difficult-to-remove surfaces. The thick crepe paper backing contains a solvent-resistant saturant that helps resist paint bleed-through against latex-based and oil-based paints. Rubber adhesive provides instant adhesion and resists lifting or curling. Tape is easy to tear, resists slivering and provides one-piece removal. Use also for holding, protecting bundling and a vast number of other difficult jobs where a pressure-sensitive tape is needed.
Catalog No. 50-008-29172 Supplier Enterprise Technology Solutions Supplier No. MMM2322
High-performance paper masking tape produces sharp paint lines in medium-temperature, paint bake operations and provides clean removal, even on difficult-to-remove surfaces. The thick crepe paper backing contains a solvent-resistant saturant that helps resist paint bleed-through against latex-based and oil-based paints. Rubber adhesive provides instant adhesion and resists lifting or curling. Tape is easy to tear, resists slivering and provides one-piece removal. Use also for holding, protecting bundling and a vast number of other difficult jobs where a pressure-sensitive tape is needed.