Learn More
3M™ PAPR Battery Charger Cradle
Catalog No. 19039566 Shop All 3M Products
Change view
Click to view available options
Battery charger cradle
Catalog No.
19-039-566 Battery charger cradle
1 options
Catalog No. 19-039-566 Supplier 3M™ Supplier No. 7000127710
TR-340 Battery Charger Cradle provides an easy-to-use interface between all TR-300 batteries and all TR-900 Series power supplies.
  • Replacement part for the TR-341 and TR-344 battery charger kits
  • 3M Catalog# TR-340
TRUSTED_SUSTAINABILITY
Specifications
Product Type Single-Unit Charging Station
For Use With (Application) Provides easy-to-use interface between all TR-300 batteries and all TR-900 series power supplies
For Use With (Equipment) Versaflo™ TR-300 and Speedglas™ TR-300 PAPR Systems
Model TR-340
No. of Ports One
Quantity 1 Case