Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Versaflo™ TR-630 PAPR Battery Pack

Catalog No. 18037 Shop All 3M Products
Change view
Click to view available options
Model 007-00-15R01
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
18-037 Model 007-00-15R01
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-037 Supplier 3M™ Supplier No. 0070015R01

missing translation for 'validMainframeMsg'

Choice of batteries used to provide power for PAPR systems

  • Rechargeable or standard batteries
  • Choice of NiMH, NiCd, Lithium or Lithium Ion
  • Long run time for filters and cartridges

These products must be used in accordance with the user instructions, warnings and limitations accompanying each product.

Compliance

NIOSH approved.

TRUSTED_SUSTAINABILITY

Specifications

ANSI Standard ANSI Z88.2-1992
Certifications (CSA) CSA Z94.4
Certifications (OSHA) OSHA, 29 CFR 1910.1200
Color Black
Product Type Battery Pack
For Use With (Application) Supplying power for PAPR systems
For Use With (Equipment) Air-Mate PAPRs
Model 007-00-15R01
Quantity 1 Ea.
Recharge Time 16 to 24 hr.
Weight (English) 1.7 lb.
Weight (Metric) 0.8 kg
Voltage 100 V
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.