Learn More
3M™ Versaflo™ TR-630 PAPR Battery Pack
Catalog No. 19813771 Shop All 3M Products
Change view
Click to view available options
Model GVP-111
Catalog No.
19-813-771 Model GVP-111
1 options
Catalog No. 19-813-771 Supplier 3M™ Supplier No. 7000029616
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Choice of batteries used to provide power for PAPR systems
  • Rechargeable or standard batteries
  • Choice of NiMH, NiCd, Lithium or Lithium Ion
  • Long run time for filters and cartridges
These products must be used in accordance with the user instructions, warnings and limitations accompanying each product.
Compliance
NIOSH approved.
TRUSTED_SUSTAINABILITY
Specifications
ANSI Standard ANSI Z88.2-1992
Certifications (CSA) CSA Z94.4
Certifications (OSHA) OSHA, 29 CFR 1910.1200
Product Type Battery Pack
For Use With (Application) Supplying power for PAPR systems
For Use With (Equipment) Breathe Easy PAPRs
Model GVP-111
Quantity 1 Case