Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ TR-300 and TR-600 PAPR Battery Packs

Catalog No. 19042682 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
Breathe Easy™ PAPR Systems
Versaflo TR-300 PAPR Series
Versaflo TR-600 PAPR Series
Color:
Black
Blue
Blue, Gray
Battery Life:
4 to 6 hr.
8 to 12 hr.
8 to 18 hr.
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Color Battery Life
19-042-682 Breathe Easy™ PAPR Systems - -
19-168-228 Breathe Easy™ PAPR Systems Blue -
19-268-020 Versaflo TR-600 PAPR Series Blue, Gray -
19-268-022 Versaflo TR-600 PAPR Series Blue, Gray 8 to 18 hr.
19-039-564 Versaflo TR-300 PAPR Series Black 4 to 6 hr.
19-039-565 Versaflo TR-300 PAPR Series Black 8 to 12 hr.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19-042-682 Supplier 3M™ Supplier No. 7000131716
Only null left
Add to Cart
Add to Cart

Choose from a selection of lithium ion rechargeable batteries for the TR-300 or TR-600 PAPR systems.

  • Rechargeable or standard batteries
  • Choice of NiCd or Lithium
  • Long run time for filters and cartridges

Battery packs must be used in accordance with the user instructions, warnings and limitations accompanying each product

Compliance

NIOSH approved.

TRUSTED_SUSTAINABILITY

Specifications

Battery Type Lithium
Certifications/Compliance NIOSH Approved
Disposable Single-use
Product Type Battery Pack
For Use With (Equipment) Breathe Easy™ PAPR Systems
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.