Learn More
Enterprise Technology Solutions 3M Particulate Respirator, with Exhalation Valve, 10/BX, White
SDP
Catalog No. 5000828111
Encompass_Preferred
Particulate respirator features 3M Cool Flow Valve technology for heat reduction and easier breathing. Advanced filter media also promotes easier breathing. Use for nonharmful dusts from sanding, grinding, sawing and insulating particles. M-nose clip allows fewer pressure points and greater comfort. Protection Paint Sanding Valved Respirator is NIOSH approved: N95.
Catalog No. 50-008-28111 Supplier Enterprise Technology Solutions Supplier No. MMM8511PB1A
Particulate respirator features 3M Cool Flow Valve technology for heat reduction and easier breathing. Advanced filter media also promotes easier breathing. Use for nonharmful dusts from sanding, grinding, sawing and insulating particles. M-nose clip allows fewer pressure points and greater comfort. Protection Paint Sanding Valved Respirator is NIOSH approved: N95.