Learn More
3M™ Peltor™ Accessories for Motorola Radios
Catalog No. 191501583 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
Motorola XTS Series Radios, Wolfpak Headset
Catalog No. For Use With (Equipment)
19-150-1583 Motorola XTS Series Radios, Wolfpak Headset
1 options
Catalog No. 19-150-1583 Supplier 3M™ Supplier No. 7010315212
3M™ Peltor™ Push-to-talk (PTT) adapter for Motorola XTS Series Radios - NATO Wired
Push-to-talk (PTT) adapter for Motorola Radios: PRC 153 IISR, Motorola XTS Series 2500;3000;3500;5000, HT1000, MT2000, MTS2000, MTX838, 2000;8000;9000, JT1000 Astro Digital - NATO Wired
  • Use 3M™ PELTOR™ Push-To-Talk (PTT) Adapters to optimize the placement of the PTT button for convenient activation
  • For use with 3M™ PELTOR™ COMTAC™™ Protective Communication Headsets and compatible two-way radios
  • Ambidextrous design allows for easy use by either the right or left hand
  • Mounting clip can be rotated 360 degrees
  • Design allows for either belt or shoulder mounting
  • Button profile and tactile feedback suitable for gloved hands
  • Optional shroud protects PTT button from accidental activation, for instance when laying in the prone position
  • NEXUS TP-120 connection serves as a break-away connector for removal in emergency situations
  • Curly cable downlead with radio specific connector To determine the appropriate PTT adapter for your two-way radio, please visit 3Mcom/PeltorAdapters
  • NATO wired
  • 3M Catalog# FL5018-02
TRUSTED_SUSTAINABILITY
Specifications
Product Type Push-to-Talk Adaptor
For Use With (Equipment) Motorola XTS Series Radios, Wolfpak Headset
Includes 30 in. Straight Cable
Model FL5018-02
Certifications/Compliance IP65