Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ PELTOR™ CH-3 Headset

PELTOR™ CH-3 Headsets are intended to be plugged into an external PTT adapter that allows for convenient placement of the Push-To-Talk button, or can also be connected to a vehicle intercom system.

Specifications

Color Bright Yellow
ANSI S3.1974 Yes
Cushion Color Black
For Use With (Application) Manufacturing
Pairs per Package 1 Ea.
View More Specs

Products 3
Catalog Number Mfr. No. Model Style Noise Reduction Rating (NRR) Price Quantity & Availability  
Catalog Number Mfr. No. Model Style Noise Reduction Rating (NRR) Price Quantity & Availability  
19-072-281
Accessories View Documents
3M™
7100203637
MT74H52A-110 Over-the-head 26 dB
Case of 10 for $2,354.15
N/A
Due to product restrictions, please to purchase or view availability for this product.
19-072-289
Accessories View Documents
3M™
7100203636
MT74H52B-110 Neckband 24 dB
Case of 10 for $2,712.19
N/A
Due to product restrictions, please to purchase or view availability for this product.
19-072-291
Accessories View Documents
3M™
7100203635
MT74H52P3E-110 Hard Hat 24 dB
Case of 10 for $2,396.15
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description

Description

  • Protective communication solution for hands-free, clear communication in noisy environments
  • FLX2 connection jack for easy and reliable connection to a wide assortment of handheld devices
  • Lasting comfort for long duration wear times
  • Designed to withstand harsh environmental elements
  • Water and dust-proof noise-canceling microphone (IP68) for clear speech transmission in noisy and harsh environments
  • Audio speakers are optimized for speech frequencies and meet the efficiency demands of many of today's digital devices
  • Bright yellow color for enhanced visibility

Compliance

3M™ ear muffs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.

Specifications

Specifications

Bright Yellow
Black
1 Ea.
No
Yes
Manufacturing
CH-3 Headset
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.