Learn More
3M™ PELTOR™ Welding Ear Muff, Behind-the-Head
Catalog No. 19121403 Shop All 3M Products
Change view
Click to view available options
Noise Reduction Rating (NRR):
17 dB
Catalog No. Noise Reduction Rating (NRR)
19-121-403 17 dB
1 options
Catalog No. 19-121-403 Supplier 3M™ Supplier No. 7000103840
Add to Cart
Add to Cart
Designed to deliver comfortable hearing protection in a wide range of welding environments, PELTOR™ behind-the-head ear muffs are for use with 3M™ welding helmets.
  • H505B ear muff low-profile design fits comfortably under most welding helmets particularly 3M™ Speedglas™ 9100 Series
  • Wire headband offers comfortability during prolonged use
  • Ear cups tilt to ensure customized fit
  • Two-point suspension for balanced pressure distribution
  • Semi-soft cup surface minimizes scratch sound transmission
Compliance
3M™ products must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
TRUSTED_SUSTAINABILITY
Specifications
ANSI S3.1974 Yes
CSA Z94.2 Class B
Cushion Color Black
For Use With (Equipment) 3 M™ Speedglas™ 9100 Series and Other Welding Helmets
Headband Material Wire
Headband Cushion No
For Use With (Application) Grinding, Machine Operations, Welding
Model H505B
Noise Reduction Rating (NRR) 17 dB
Headband Color Silver
Dielectric No
Style Behind-the-head