Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ PELTOR™ Welding Hygiene Kits

Catalog No. 19121400 Shop All 3M Products
Change view
Click to view available options
Cushion Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Cushion Color
19-121-400 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-121-400 Supplier 3M™ Supplier No. 7000103822
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PELTOR™ hygiene kit contains replacement cushions and inserts for the H505B welding ear muff.

  • Hygiene replacement kit for the H505B welding ear muff
  • Contains cushions and inserts- 10 each/cs.
  • Recommended replacement is every six months or whenever they become damaged

Compliance

3M™ products must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.

TRUSTED_SUSTAINABILITY

Specifications

Cushion Color Black
For Use With (Equipment) Hygiene Kit for H505B Welding Ear Muff
Model HY505
Product Type Hygiene Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.