Learn More
3M™ PELTOR™ X Series Ear Muff Probed Test Cushions
Catalog No. 19122404 Shop All 3M Products
Change view
Click to view available options
Model:
393-3001-2
393-3003-2
393-3005-2
For Use With (Equipment):
X Series X1 and X2 Ear Muffs
X Series X3 Ear Muffs
X Series X4 and X5 Ear Muffs
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model For Use With (Equipment)
19-122-404 393-3001-2 X Series X1 and X2 Ear Muffs
19-122-406 393-3005-2 X Series X4 and X5 Ear Muffs
19-122-405 393-3003-2 X Series X3 Ear Muffs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-122-404 Supplier 3M™ Supplier No. 7100062126
Add to Cart
Add to Cart
Probed Test Cushions easily snap into the 3M™ PELTOR™ X Series Ear Muffs (sold separately) to allow for fit testing with the E-A-Rfit™ Dual-Ear Validation System.
  • Easily snaps into employee's own ear muffs (sold separately) to enable fit test
  • For use with 3M™ PELTOR™ X Series ear muffs and the E-A-Rfit™ dual-ear validation system
  • Probed cushions can be cleaned with hygiene wipes and reused
  • Adapter clips hold microphones next to ear muffs during testing
Compliance
3M™ products must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
TRUSTED_SUSTAINABILITY
Specifications
Cushion Color Black
For Use With (Equipment) X Series X1 and X2 Ear Muffs
For Use With (Application) Fit testing with the 3M™ E-A-Rfit™ dual ear validation system
Model 393-3001-2
Product Type Probed Ear Muff Test Cushions
Material PVC