Learn More
3M™ PELTOR™ Hygiene Kit for X Series Ear Muffs
Catalog No. 17000626 Shop All 3M Products
Change view
Click to view available options
Model:
HYX1
HYX2
HYX5
For Use With (Equipment):
PELTOR™ X1 Ear Muffs
PELTOR™ X2 Ear Muffs
PELTOR™ X5 Ear Muffs
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model For Use With (Equipment)
17-000-626 HYX1 PELTOR™ X1 Ear Muffs
17-000-627 HYX2 PELTOR™ X2 Ear Muffs
17-000-630 HYX5 PELTOR™ X5 Ear Muffs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17-000-626 Supplier 3M™ Supplier No. 7000104045
Add to Cart
Add to Cart
3M™ PELTOR™ Hygiene Kit includes replaceable ear cushions and foam liners for X Series Ear Muffs
  • Hygiene kits consist of one pair of replacement cushions and foam inserts for 3M™ PELTOR™ X Series ear muffs (X1, X2, X3, X4, X5)
  • 3M recommends replacing ear muff cushions and foam inserts every six months or whenever they become damaged.
  • Kits are packaged 10 Each/Cs.
Compliance
3M™ products must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
TRUSTED_SUSTAINABILITY
Specifications
Cushion Color Black
For Use With (Equipment) PELTOR™ X1 Ear Muffs
For Use With (Application) Assembly, Blasting, Cleaning, Demolition, Electrical, Facility Maintenance, Grinding, Machine Operations, Painting, Sanding, Welding
Model HYX1
Product Type Hygiene Kit
Dielectric Yes