Learn More
3M™ Peltor™ In-Line Push-To-Talk Adaptor
Catalog No. 191501590 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
Ericsson Jaguar 700P Nato-DC Series
Catalog No. For Use With (Equipment)
19-150-1590 Ericsson Jaguar 700P Nato-DC Series
1 options
Catalog No. 19-150-1590 Supplier 3M™ Supplier No. 7010341813
In-Line Push-To-Talk Adaptor FL5083-02, Ericsson Jaguar 700P Nato-DC 1 EA/Case
Use 3M™ PELTOR™ Push-To-Talk (PTT) Adapters to optimize the placement of the PTT button for convenient activation.
  • For use with 3M™ PELTOR™ COMTAC™™ Protective Communication Headsets and compatible two-way radios
  • Ambidextrous design allows for easy use by either the right or left hand
  • Mounting clip can be rotated 360 degrees
  • Design allows for either belt or shoulder mounting
  • Button profile and tactile feedback suitable for gloved hands
  • Optional shroud protects PTT button from accidental activation, for instance when laying in the prone position
  • NEXUS TP-120 connection serves as a break-away connector for removal in emergency situations
  • Curly cable downlead with radio specific connector To determine the appropriate PTT adapter for your two-way radio, please visit 3Mcom/PeltorAdapters
  • NATO wired
  • 3M Catalog# FL5083-02
TRUSTED_SUSTAINABILITY
Specifications
Product Type Push-to-Talk Adaptor
For Use With (Equipment) Ericsson Jaguar 700P Nato-DC Series
Model FL5083-02
Certifications/Compliance IP65