Learn More
3M™ PELTOR™ Optime™ 101 Ear Muffs
Catalog No. 19060609 Shop All 3M Products
Change view
Click to view available options
Model:
H7A
H7B
H7P3E
Style:
Behind-the-head
Hard Hat
Over-the-head
Noise Reduction Rating (NRR):
24 dB
26 dB
27 dB
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Style Noise Reduction Rating (NRR)
19-060609 H7B Behind-the-head 26 dB
17-388-1A H7A Over-the-head 27 dB
17-388-1B H7P3E Hard Hat 24 dB
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-060609 Supplier 3M™ Supplier No. 7000002329
Add to Cart
Add to Cart
This item is not returnable. View return policy
PELTOR™ Optime™ 101 Ear Muffs are recommended for TWA noise exposures up to 101 dBA.
  • For noise exposures up to 101 dBA
  • Liquid-foam ear cushion
  • Ear cushions snap in and out of place for easy cleaning
  • Over-the-Head Style or Hard Hat-Attached styles
  • Adjusts to fit any head size
  • Soft, padded headband for extra comfort
  • Stainless-steel band wires
  • Four-point suspension system distributes suspension evenly
Compliance
3M™ ear muffs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
TRUSTED_SUSTAINABILITY
Specifications
Color Black, Green
ANSI S3.1974 Yes
Cushion Material Liquid-foam
CSA Z94.2 Class A
Cushion Color Black
For Use With (Application) Assembly, cleaning, demolition, electrical, facility maintenance, grinding, machine operations, painting, sanding, welding
High Visibility No
Model H7B
Pairs per Package 1 Ea.
Noise Reduction Rating (NRR) 26 dB
Product Type PELTOR™ Optime™
Dielectric No
Material ABS
Style Behind-the-head