Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ PELTOR™ Optime™ 105 Ear Muffs

Catalog No. 191301978 Shop All 3M Products
Change view
Click to view available options
Model:
H10A
H10AHV
H10B
H10P3E
Color:
Black, Red
High-visibility Lime
Red
Style:
Hard Hat
Neckband
Over-the-head
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Color Style
19-130-1978 H10AHV High-visibility Lime Over-the-head
18-858N H10B Black, Red Neckband
19-039-190 H10P3E Black, Red Hard Hat
18-858M H10A Red Over-the-head
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-130-1978 Supplier 3M™ Supplier No. 7100008764
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Optime™ Ear Muffs provide maximum sound quality in the most demanding environments where noise exposures are expected to reach as high as 105 dBA.

  • Effective hearing protection for noise exposures up to 105dBA
  • Double-shell ear cup design technology boosts noise blocking ability
  • Soft, foam-filled ear cushions provide added comfort
  • Durable stainless steel headband resists bending and warping
  • Available in headband style red/black or high-visibility green color
  • Available in neckband and hard hat-attached styles

Compliance

3M™ ear muffs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.

TRUSTED_SUSTAINABILITY

Specifications

Color High-visibility Lime
ANSI S3.1974 Yes
Cushion Material Foam
CSA Z94.2 Class AL
Cushion Color Black
Headband Material Stainless Steel
For Use With (Application) Assembly, cleaning, demolition, facility maintenance, grinding, machine operations, painting, sanding, welding
Model H10AHV
Pairs per Package 1 Ea.
Noise Reduction Rating (NRR) 30 dB
Product Type PELTOR™ Optime 105
Headband Color Black
Dielectric No
Material ABS
Style Over-the-head
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.