Learn More
Nelson Jameson Company 3M Petrifilm Yeast and Mold Count Plates, 1000 plates/case, sample-ready culture medium system, compatible with standard colony counters, 20-25°C incubation
Catalog No. NC2060780
Encompass
3M Petrifilm Yeast and Mold Count Plates, 1000 plates/case, sample-ready culture medium system, compatible with standard colony counters, 20-25°C incubation
Catalog No. NC2060780 Supplier Nelson Jameson Company Supplier No. 4603417 C
3M Petrifilm Yeast and Mold Count Plates, 1000 plates/case, sample-ready culture medium system, compatible with standard colony counters, 20-25°C incubation