Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Petroleum Pillows

Catalog No. 19039456 Shop All 3M Products
Change view
Click to view available options
Capacity (English):
3.5 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
19-039456 3.5 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-039456 Supplier 3M™ Supplier No. 7100003891
Add to Cart
Add to Cart

  • 3M Catalog# T240

TRUSTED_SUSTAINABILITY

Specifications

Capacity (English) 3.5 gal.
Capacity (Metric) 13.24 L
Type Petroleum Sorbent Pillow
For Use With (Application) Hazardous liquid spills
Model T240
Dimensions (L x W) 22 x 18 in. (55.8 x 45.72 cm)
Length (English) 22 in.
Length (Metric) 55.8 cm
Width (English) 18 in.
Width (Metric) 45.72 cm
Weight (English) 2.4 lb.
Weight (Metric) 1.08 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.