Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Petroleum Sorbents, T-Series

Catalog No. 19039465 Shop All 3M Products
Change view
Click to view available options
Capacity (English):
10 gal.
12 gal.
Diameter (English):
3 in.
4 in.
Length (English):
32 in.
8 ft.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Diameter (English) Length (English)
19-039465 10 gal. 4 in. 32 in.
19-039461 12 gal. 3 in. 8 ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
  • Failure to properly handle and dispose of sorbents contaminated by hazardous materials can result in serious physical injury or death and/or environmental harm.
  • Sorbents may become slippery when used on flooring. Do not walk on sorbents or place in high traffic areas
Catalog No. 19-039465 Supplier 3M™ Supplier No. 7000006449
Add to Cart
Add to Cart
  • Failure to properly handle and dispose of sorbents contaminated by hazardous materials can result in serious physical injury or death and/or environmental harm.
  • Sorbents may become slippery when used on flooring. Do not walk on sorbents or place in high traffic areas

Petroleum sorbent mini-booms are used to encircle spills and to contain leaks and are a selective sorbent which will sorb hydrocarbon-based liquids (oleophilic) and repel aqueous-based liquids (hydrophobic)

  • For use in oil-only sorbent applications
  • Available in three lengths.
  • Ideal for placing around equipment.
  • Helps prevent leaks/spills from spreading to adjacent areas.
  • Scrim on the outside of mini-boom identifies the use while containing particulate sorbent material
  • 3M Catalog# T280 , 3M Catalog# T8

TRUSTED_SUSTAINABILITY

Specifications

Capacity (English) 10 gal.
Capacity (Metric) 37.85 L
Type Petroleum Sorbent Double Boom
For Use With (Application) Used to encircle spills and to contain leaks
Model T280
Dimensions (L x W) 32 x 20 in. (81.3 x 50.8 cm)
Length (English) 32 in.
Length (Metric) 81.3 cm
Width (English) 20 in.
Width (Metric) 50.8 cm
Diameter (English) 4 in.
Diameter (Metric) 10.16 cm
Weight (English) 6.5 lb.
Weight (Metric) 2.94 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.