Learn More
Kd Medical Inc 3M POTASSIUM ACETATE BOTTLE
SDP
Catalog No. 501018899
Encompass_Preferred
3M Potassium Acetate Solution, pH5.2, is used for preparation of BAC and PAC DNA. This solution is a sterile stock. One 100ml bottle.
Catalog No. 50-101-8899 Supplier Kd Medical Inc Supplier No. RGC5220
3M Potassium Acetate Solution, pH5.2, is used for preparation of BAC and PAC DNA. This solution is a sterile stock. One 100ml bottle.