Learn More
Enterprise Technology Solutions 3M Power Sponge,Dual Pad/Sponge,4-1/2"x2-4/5"x7/10",5/PK,AA
SDP
Catalog No. 5000829154
Encompass_Preferred
Power Sponges offer a dual-action cleaning pad and sponge in one to clean many surfaces. Unique bow-tie shape for easy handling. The sponge side contains antimicrobial protection to resist odors, staining and crumbling caused by bacteria. Scouring side of the product scours two-to-four times faster and is six-to-ten times less abrasive than other conventional commercial scouring pads to reduce scratching. Unique, soil-removing particles deliver effective scouring, cleaning and scrubbing. Particles are evenly distributed throughout the scouring pad to ensure effective, consistent scouring and lasting performance throughout the life of the pad. Durable synthetic fibers are crimped and heat-set to create strong web. High-quality bonding resin system resists degradation from exposure to hot water, detergents and normal cleaning liquids
Catalog No. 50-008-29154 Supplier Enterprise Technology Solutions Supplier No. MMM3000CC
Power Sponges offer a dual-action cleaning pad and sponge in one to clean many surfaces. Unique bow-tie shape for easy handling. The sponge side contains antimicrobial protection to resist odors, staining and crumbling caused by bacteria. Scouring side of the product scours two-to-four times faster and is six-to-ten times less abrasive than other conventional commercial scouring pads to reduce scratching. Unique, soil-removing particles deliver effective scouring, cleaning and scrubbing. Particles are evenly distributed throughout the scouring pad to ensure effective, consistent scouring and lasting performance throughout the life of the pad. Durable synthetic fibers are crimped and heat-set to create strong web. High-quality bonding resin system resists degradation from exposure to hot water, detergents and normal cleaning liquids