Learn More
Enterprise Technology Solutions 3M Prep Pad, f/Finish Removal, 14"x20", 10/CT, Red
SDP
Catalog No. 5000790622
Encompass_Preferred
Scotch-Brite Surface Preparation Pads require only neutral cleaner or water to effectively remove floor finish, preparing the floor for finish application. Optimized mineral abrasives leave less hazing and scratching than sanding screens. Use with rotary or automatic floor machines. High-quality nylon fibers are coated throughout the pad with resin and abrasives, providing a consistently aggressive cut. Versatile pad prepares wood, vinyl tile, sheet vinyl, marble, terrazzo and concrete floors for finish applications. It helps enhance safety and productivity - no slippery residue from stripper; no cleanup on baseboards or nearby floors; no chemical contamination on skin, clothes and shoes. Pad aggressively scrubs finishes in preparation for recoating.
Catalog No. 50-007-90622 Supplier Enterprise Technology Solutions Supplier No. MMMSPP14X20
Scotch-Brite Surface Preparation Pads require only neutral cleaner or water to effectively remove floor finish, preparing the floor for finish application. Optimized mineral abrasives leave less hazing and scratching than sanding screens. Use with rotary or automatic floor machines. High-quality nylon fibers are coated throughout the pad with resin and abrasives, providing a consistently aggressive cut. Versatile pad prepares wood, vinyl tile, sheet vinyl, marble, terrazzo and concrete floors for finish applications. It helps enhance safety and productivity - no slippery residue from stripper; no cleanup on baseboards or nearby floors; no chemical contamination on skin, clothes and shoes. Pad aggressively scrubs finishes in preparation for recoating.