Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ W-3217 Vinyl Waist Belt

Catalog No. 18183 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
W-Series and V-Series
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
18-183 W-Series and V-Series
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-183 Supplier 3M™ Supplier No. 7000130997

missing translation for 'validMainframeMsg'

For use with W-Series and V-Series air control valves.

  • These safety products must be used in accordance with the user instructions, warnings and limitations accompanying each product.
  • 3M Catalog# W-3217

TRUSTED_SUSTAINABILITY

Specifications

Color Black
Disposable Reusable
Material Vinyl
Product Type Belt
For Use With (Application) Keeps supplied air hose in place
For Use With (Equipment) W-Series and V-Series
Model W-3217
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.