Learn More
3M™ Privo™ Safety Eyewear
Catalog No. 19166813 Shop All 3M Products
Change view
Click to view available options
Frame Color:
Black
Silver
Lens Tint:
Amber
Clear
Gray
Indoor/Outdoor Mirror
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Tint
19-166-813 Black Indoor/Outdoor Mirror
19-166-810 Black Clear
19-166-812 Black Amber
19-166-814 Silver Clear
19-166-815 Silver Gray
19-166-817 Silver Indoor/Outdoor Mirror
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Catalog No. 19-166-813 Supplier 3M™ Supplier No. 7000127537
Add to Cart
Add to Cart
This item is not returnable. View return policy
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Offers European styling
  • Lens and temple tip color options
  • Soft-tipped temples
  • Soft comfort nose bridge
  • Durable polycarbonate lenses absorb 99.9% UV
  • 3M Catalog# 12261-00000-20, 12263-00000-20, 12264-00000-20, 12265-00000-20, 12266-00000-20, 12268-00000-20
Compliance
Meets ANSI™ Z87.1-2010 and CSA Z94.3 standards.
TRUSTED_SUSTAINABILITY
Specifications
Lens Type I/O Mirror
Certifications/Compliance Meets ANSI Z87.1-2010 and CSA Z94.3 standards.
Lens Tint Indoor/Outdoor Mirror
Frame Color Black
Model 12264-00000-20
Temple Color Orange
Lens Material Polycarbonate