Learn More
Enterprise Technology Solutions 3M Privo Safety Glass, Orange/Black
SDP
Catalog No. 5000828478
Encompass_Preferred
Stylish Privo unisex protective eyewear looks as smooth as it functions. Half-frame design bridges Euro-style and comfort with soft tips and temples, a gentle nose bridge, and a sturdy polycarbonate lens that absorbs 99.9 percent ultraviolet light. Anti-fog coating allows all-day, indoor or outdoor use. Recommended for assembly, cleaning, electrical, facility maintenance, grinding, machine operations, painting and sanding, these safety glasses help protect your eyes from impact and ultraviolet light. Safety glasses meet the High Impact Requirements of ANSI Z87.1-2003.
Catalog No. 50-008-28478 Supplier Enterprise Technology Solutions Supplier No. MMM122610000020
Stylish Privo unisex protective eyewear looks as smooth as it functions. Half-frame design bridges Euro-style and comfort with soft tips and temples, a gentle nose bridge, and a sturdy polycarbonate lens that absorbs 99.9 percent ultraviolet light. Anti-fog coating allows all-day, indoor or outdoor use. Recommended for assembly, cleaning, electrical, facility maintenance, grinding, machine operations, painting and sanding, these safety glasses help protect your eyes from impact and ultraviolet light. Safety glasses meet the High Impact Requirements of ANSI Z87.1-2003.