Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Probed Particulate Respirator, P100

Catalog No. 19813738 Shop All 3M Products
Change view
Click to view available options
Standard size
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-813-738 Standard size
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-813-738 Supplier 3M™ Supplier No. 7010383764

Quantitative fit test respirator only. Must not be used for respiratory protection.

  • Respirator is probed and used for quantitative fit testing ONLY
  • Not to be used for protection in a contaminated area
  • 3M Catalog# 8293Q
TRUSTED_SUSTAINABILITY

Specifications

Product Type Quantitative Fit Test Particulate Respirator
Filter Type P100
With Exhalation Valve Yes
Aerosol Type P - Oil-Proof
Color White
Design Type Cup-shaped
FDA Cleared Surgical Mask No
Filter Efficiency 0.9997
For Use With (Application) Quantitative Fit Testing
Latex Free Yes
Material Polypropylene, Polyester
Model 8293Q
Nosepiece Type Nose clip
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.