Learn More
Enterprise Technology Solutions 3M Protective Eyewear, Anti-Fog Lens, 20/Case, Gray
SDP
Catalog No. 5000827961
Encompass_Preferred
SecureFit Protective Eyewear features pressure diffusion technology that naturally adjusts to individual head sizes for a custom, natural fit as soon as you put them on. The self-adjusting construction adapts to each wearer and provides a snug fit that keeps the glasses securely in place as the wearer looks up and down or side-to-side to eliminate the need for frequent repositioning. Universal design eliminates the expense of stocking multiple frame sizes at jobsites to accommodate different people to keep your costs to a minimum. Impact-resistant polycarbonate lenses absorb 99.9 percent UVA and UVB rays up to 380 nm to maintain healthy vision when used outdoors. Anti-fogging properties help maintain visibility in hot and humid environments to help increase usage on the job.
Catalog No. 50-008-27961 Supplier Enterprise Technology Solutions Supplier No. MMMSF202AF
SecureFit Protective Eyewear features pressure diffusion technology that naturally adjusts to individual head sizes for a custom, natural fit as soon as you put them on. The self-adjusting construction adapts to each wearer and provides a snug fit that keeps the glasses securely in place as the wearer looks up and down or side-to-side to eliminate the need for frequent repositioning. Universal design eliminates the expense of stocking multiple frame sizes at jobsites to accommodate different people to keep your costs to a minimum. Impact-resistant polycarbonate lenses absorb 99.9 percent UVA and UVB rays up to 380 nm to maintain healthy vision when used outdoors. Anti-fogging properties help maintain visibility in hot and humid environments to help increase usage on the job.