Learn More
Enterprise Technology Solutions 3M Protective Eyewear, Wraparound, Adj.Headband, BK/Clear
SDP
Catalog No. 5000828113
Encompass_Preferred
Protection Professional Chemical Splash/Impact Goggles help protect eyes from certain airborne particulates and chemical splash. They are constructed with a polycarbonate, wraparound, oversized lens that fits over most personal prescription eyewear, and is .060 thick. They also provide excellent peripheral vision and have an adjustable, chemical-resistant and flame-resistant headband. Vents are designed to help prevent liquids from entering the eye area. These goggles provide lightweight, comfortable protection. They have anti-fog lens and meet requirements of ANSI Z87.1-2003.
Catalog No. 50-008-28113 Supplier Enterprise Technology Solutions Supplier No. MMM91264H1DC
Protection Professional Chemical Splash/Impact Goggles help protect eyes from certain airborne particulates and chemical splash. They are constructed with a polycarbonate, wraparound, oversized lens that fits over most personal prescription eyewear, and is .060 thick. They also provide excellent peripheral vision and have an adjustable, chemical-resistant and flame-resistant headband. Vents are designed to help prevent liquids from entering the eye area. These goggles provide lightweight, comfortable protection. They have anti-fog lens and meet requirements of ANSI Z87.1-2003.