Description
- Suspension allows to customize height and fit
- Vents allow air circulation to keep worker cooler and more comfortable
- Meets requirements of ANSI/ISEA Z89.1-2009 Type 1, Class C
- ANSI/ISEA compliant when worn in the forward and reverse donning positions
- Replaceable brow pad for comfort and extends service life
- Short brim for upward visibility
- Available in various colors
- 3M Catalog# H-701V, H-702V, H-703V, H-704V, H-711V, H-712V, H-713V
Specifications
Specifications
| Certifications/Compliance | ANSI/ISEA Z89.1-2014, Type 1 |
| Color | Green |
| Includes | Standard brow pad |
| Model | H-704V |
| Product Type | Cap-style Hard Hat |
| Quantity | 20/Cs. |
| Suspension Type | 4-Point Ratchet |
| Ventilated | Ventilated |