Learn More
Nelson Jameson Company 3M Quick Swabs, 50/pk
Catalog No. NC1161009
Encompass
This ready-to-use environmental swab system consists of a 5-inch rayon-tipped swab which contains letheen neutralizing buffer to facilitate recovery of bacteria. The swabs can be used wet or dry to sample surfaces and quickly deliver 1.0 mL of sample onto a 3M Petrifilm Plate. Handy vinyl-covered wire rack which holds up to 50 swabs is sold separately.
Catalog No. NC1161009 Supplier Nelson Jameson Company Supplier No. 4603432 C
This ready-to-use environmental swab system consists of a 5-inch rayon-tipped swab which contains letheen neutralizing buffer to facilitate recovery of bacteria. The swabs can be used wet or dry to sample surfaces and quickly deliver 1.0 mL of sample onto a 3M Petrifilm Plate. Handy vinyl-covered wire rack which holds up to 50 swabs is sold separately.